Learn More
National Safety Apparel™ Women's 12 Cal UltraSoft FR Coveralls 30 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$228.00 - $303.00
Specifications
Material Cotton, Nylon
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-581
View Documents
National Safety Apparel™
C88UPWSM30
Small
Each for $228.00
Add to cart
17-001-582
View Documents
National Safety Apparel™
C88UPWMD30
Medium
Each for $228.00
Add to cart
17-001-583
View Documents
National Safety Apparel™
C88UPWLG30
Large
Each for $228.00
Add to cart
17-001-584
View Documents
National Safety Apparel™
C88UPWXL30
X-Large
Each for $228.00
Add to cart
17-001-585
View Documents
National Safety Apparel™
C88UPW2X30
2X-Large
Each for $251.00
Add to cart
17-001-586
View Documents
National Safety Apparel™
C88UPW3X30
3X-Large
Each for $251.00
Add to cart
17-001-587
View Documents
National Safety Apparel™
C88UPW4X30
4X-Large
Each for $276.00
Add to cart
17-001-588
View Documents
National Safety Apparel™
C88UPW5X30
5X-Large
Each for $303.00
Add to cart
Description
Description
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps
  • Home laundry
  • Made in the USA
Specifications
Specifications
Cotton, Nylon
Stitched