Learn More
Fisherbrand™ Wood Adjustable Height Table, 72 in. Wide, Black Epoxy Work Surface
Fisherbrand™ Wood Adjustable Height Tables bring quality, warmth and durability to your laboratory.
Description
- Made from hand selected premium rotary cut maple or rift cut oak veneers.
- Robotically applied environmentally friendly finishes are chemically resistant.
- Wood Adjustable Height Tables have a structural steel upper frame that is wrapped with wood rails for maximum strength.
- The tables are adjustable in height via a stainless steel insert with in the 2.25 in. wooden legs
- They are adjustable in 1 in. increments, from 30 to 36 in. and include nylon leveling feet.
- They are desgiend to accommodate the wood mobile cabinets in the Fisherbrand Program
Compliance
SEFA 8
Specifications
Honey Stain | |
76.2 cm | |
72 in. | |
Table Frame, Epoxy Work Surface, Stainless Steel Adjustable Insert, Nylon Leveling Glides | |
182.9 cm |
76.2 to 91.4 cm | |
30 to 36 in. | |
30 in. | |
2 years | |
Maple Veneer |