Learn More
Fisherbrand™ ADA Height Rear and Center Wood Fillers
Fisherbrand™ Rear and Center Fillers bring quality, warmth and durability to your laboratory.
Description
- Made from hand selected premium rotary cut maple or rift cut oak veneers
- Robotically applied environmentally friendly finishes are chemically resistant.
- Rear and Center Fillers are used between back-to-back base cabinets or to close space at the rear of a cabinet.
- Fillers can be cut to fit in the field
Compliance
SEFA 8
Specifications
Amber Stain | |
1 in. | |
32.5 in. | |
2 to 18 in. |
Wood (Oak Veneer) | |
2.5 cm | |
82.6 cm | |
5.1 to 45.7 cm |