Learn More
Fisherbrand Wooden Splints
Thin wood
This item is not returnable.
View return policy
Description
Sterile are individually wrapped: 1/envelope, 100 envelopes/pack, 10 packs/cs
- Sterile and non-sterile products available (see product specifications)
- Made of taste-free, splinter-free, straight-grained, smooth white birch
- For oral patient exams and care, gathering specimen, stirring and more
- Depressor is 6L x 0.75 in.W (15 x 1.9cm)
Specifications
4.5 in. | |
11.4 cm | |
Splint | |
Wood |