Learn More
Work Area Protection PVC Traffic Cones Non-distribution product as customer accommodation.
High visibility under all weather conditions
Manufacturer:  Work Area Protection 28PVCS
 View more versions of this product
Catalog No. 19-821-062
Add to cart
Specifications
Specifications
26.67cm
Fluorescent Orange
28 in.
71.1cm
7 lb.
15.25 in.
10.5 in.
Traffic Cone
PVC
0.13 in. (bottom ), 0.075 in. (top)
3.17kg
38.7cm