Learn More
Brady™ WorkHorse™ B-423 Permanent Polyester Labels
For use in commercial/industrial applications. Brady B-423 polyester label are for use with BMP71 printer.
Manufacturer:  Brady™ M714423
 View more versions of this product
Catalog No. 19-109-148
Add to cart
Specifications
Specifications
Acrylic, Permanent
UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815)
10.16 mm10.16 mm
Asset tracking, barcoding, component ID, equipment ID, general purpose ID, rating plates
10.16 mm
Square
W x H: 10.16 x 10.16
0.003 in.
13.34 mm
Polyester (B-423)
500 Labels/Roll
White
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
Gloss
BMP71 printer
1
W x H: 0.4 x 0.4 in.
-70°C to +110°C
0.076 mm
10.16 mm
Thermal Transfer portable printer label