Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-488
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of polyester and is used for thermal transfer printers.
$61.10 - $226.00
Specifications
Product Type Thermal Transfer Printable Label
Format Roll
Material Polyester (B-488)
Color White
Thickness (English) 0.0036 in.
View More Specs
Products 7
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
19-109-208 View Documents Brady™
THT1448810
W x H: 16.51 x 5.08 mm Each for $90.00
Add to cart
19-109-219 View Documents Brady™
THT154882.5
W x H: 50.8 x 6.35 mm Each for $73.80
Add to cart
19-109-230 View Documents Brady™
THT1644881
W x H: 22.86 x 22.86 mm Each for $61.10
Add to cart
19-109-269 View Documents Brady™
THT184883
W x H: 76.2 x 25.4 mm Each for $226.00
Add to cart
19-109-213 View Documents Brady™
THT148810
W x H: 19.05 x 6.35 mm Each for $90.00
Add to cart
19-109-286 View Documents Brady™
THT194881
W x H: 76.2 x 50.8 mm Each for $177.10
Add to cart
19-109-307 View Documents Brady™
THT248810
W x H: 22.86 x 6.35 mm Each for $120.00
Add to cart
Description
Description
  • Available in various sizes
Specifications
Specifications
Thermal Transfer Printable Label
Polyester (B-488)
0.0036 in.
Acrylic, Permanent
Matte
Asset Tracking, Barcode Labeling, Circuit Board Identification - Through Hole Top, Component and Equipment Identification, General and Industrial Identification, Rating Plates
Roll
White
Rectangle
Recommended ribbon series: R4300; R6200
UL Recognized; CSA Accepted
BBP™ 81 Label Printer; IP™ Series; PR Plus Series Printers