Learn More
MSA™ Workman™ Full Body Harnesses
For working in confined spaces and at heights. MSA™ Workman™ Harnesses provide optimum quality and fall arrest systems for workers.
Supplier: MSA™ 10072480
View more versions of this product
Safety Services Available
Overview
Fall Protection Assessment
Description
- Provides protection due to its lightweight components and durable webbing
- Vest-style harness with optional hip D-Rings, fits wide range of body types and sizes
- Color-contrasting thigh and torso straps simplify donning
- Econo
Specifications
X-Large | |
OSHA regulations, ANSI A10.32, ANSI Z359.1, CSA Z259.10-06 | |
Qwik-Fit chest strap | |
Working in confined spaces and at heights | |
Stainless Steel |
Polyester | |
Full body harness | |
Back | |
Qwik-Fit leg buckles | |
Yellow |