Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Workrite™ 4.5 oz Industrial Nomex™ Coveralls

Manufacturer:  Workrite™ 110NMX45OR440R

 View more versions of this product

Catalog No. 19-082356


Add to cart

Specifications

Specifications

Nomex™ IIIA
1
Orange
Industrial puposes
Stitched
Regular
Safety
Zipper with snap front
4.5 oz. Nomex™ IIIA construction
6
44 in.
Elastic
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit