Learn More
Workrite™ 4.5 oz Industrial Nomex™ Coveralls
$170.10 - $380.50
Specifications
Arc Flash Rating 1
Closure Type Zipper with snap front
Description 4.5 oz. Nomex™ IIIA construction
Material Nomex™ IIIA
No. of Pockets 6
View More Specs
Products 20
Catalog Number Mfr. No. Color Style Size (English) Chest Price Quantity & Availability  
Catalog Number Mfr. No. Color Style Size (English) Chest Price Quantity & Availability  
19-082367
View Documents
Workrite™
110NX45NB48 0S
Navy Short 48 in. Each for $344.50
Add to cart
19-082423
View Documents
Workrite™
110NMX450OR420L
Orange Long 42 in. Each for $207.50
Add to cart
19-082368
View Documents
Workrite™
110NX45NB52 0R
Navy Regular 52 in. Each for $380.50
Add to cart
19-082361
View Documents
Workrite™
110NX45NB36 0S
Navy Short 36 in. Each for $212.50
Add to cart
19-082362
View Documents
Workrite™
110NX45NB38 0S
Navy Short 38 in. Each for $170.10
Add to cart
19-082363
View Documents
Workrite™
110NX45NB40 0S
Navy Short 40 in. Each for $170.10
Add to cart
19-082365
View Documents
Workrite™
110NX45NB44 0S
Navy Short 44 in. Each for $321.00
Add to cart
19-082364
View Documents
Workrite™
110NMX45NB420S
Navy Short 42 in. Each for $170.10
Add to cart
19-087187
View Documents
Workrite™
110NMX45NB500R
Navy Regular 50 in. Each for $170.10
Add to cart
19-082366
View Documents
Workrite™
110NX45NB46 0R
Navy Regular 46 in. Each for $321.00
Add to cart
19-082353
View Documents
Workrite™
110NMX45OR360R
Orange Regular 36 in. Each for $207.50
Add to cart
19-082369
View Documents
Workrite™
110NMX45NB560R
Navy Regular 56 in. Each for $380.50
Add to cart
19-082354
View Documents
Workrite™
110NMX45OR400R
Orange Regular 40 in. Each for $207.50
Add to cart
19-082422
View Documents
Workrite™
110NMX45OR400L
Orange Long 40 in. Each for $207.50
Add to cart
19-082421
View Documents
Workrite™
110NMX45OR360L
Orange Long 36 in. Each for $207.50
Add to cart
19-082424
View Documents
Workrite™
110NMX45OR440L
Orange Long 44 in. Each for $207.50
Add to cart
19-082355
View Documents
Workrite™
112NX45ORMD 0R
Orange Regular 42 in. Each for $207.50
Add to cart
19-082356
View Documents
Workrite™
110NMX45OR440R
Orange Regular 44 in. Each for $207.50
Add to cart
19-082425
View Documents
Workrite™
110NMX45OR480L
Orange Long 48 in. Each for $207.50
Add to cart
19-082357
View Documents
Workrite™
112NX45ORXL 0R
Orange Regular 48 in. Each for $207.50
Add to cart
Specifications
Specifications
1
4.5 oz. Nomex™ IIIA construction
6
Industrial puposes
Zipper with snap front
Nomex™ IIIA
Stitched
Elastic