Learn More
Workrite™ Flame-Resistant Lab Coats
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
Lab coat with concealed snap front closure and seamed back for closure
- Made of 6 oz. Nomex™ IIIA fabric
- Four chevron pockets for ample storage
- Side pass-thru for access to inner clothing
- Knee length, worn directly over street clothes
- Color: Royal Blue
- Available in various sizes
- ARC Rating: 5.7
- HRC Category: 1
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire.
Specifications
Size | 3X-Large |
Material | Nomex IIIA |
Color | Royal Blue |