Learn More
Workrite™ Flame-Resistant Lab Coats for Women
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Made of Nomextrademark (TM) IIIA fabric
- Three pockets and lay down collar with snap
- Side Pass throughs to inner clothing
- Knee length, worn directly over street clothes
- ARC Rating: 5.7
- HRC Category: 1
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire.
Specifications
Size | Large |
Material | Nomex IIIA |
Color | Blue |