Learn More
Workrite™ Flame-Resistant Lab Coats for Women
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Fitted back with mid back belt for tapered look
  • Made of Nomextrademark (TM) IIIA fabric
  • Three pockets and lay down collar with snap
  • Side Pass throughs to inner clothing
  • Knee length, worn directly over street clothes
  • ARC Rating: 5.7
  • HRC Category: 1
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire.
Size 5X-Large
Material Nomex™ IIIA
Color Blue