Learn More
Workrite™ Flame-Resistant Lab Coats for Women
Fitted back with mid back belt for tapered look
$130.00 - $161.50
Specifications
Color Blue
Material Nomex™ IIIA
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-821-602
View Documents
Workrite™
351NX60RBLG 00
Large Each for $130.00
Add to cart
19-821-603
View Documents
Workrite™
351NX60RB XL 0R
X-Large Each for $130.00
Add to cart
19-821-600
View Documents
Workrite™
351NX60RBSM 00
Small Each for $130.00
Add to cart
19-821-601
View Documents
Workrite™
351NX60RBMD 00
Medium Each for $144.30
Add to cart
19-821-605
View Documents
Workrite™
351NX60RB3L 00
3X-Large Each for $161.50
Add to cart
19-821-604
View Documents
Workrite™
351NX60RB2L 00
2X-Large Each for $161.50
Add to cart
19-821-606
View Documents
Workrite™
351NX60RB4L 00
4X-Large Each for $161.50
Add to cart
19-821-607
View Documents
Workrite™
351NX60RB5L 00
5X-Large Each for $161.50
Add to cart
Description
Description
  • Made of Nomextrademark (TM) IIIA fabric
  • Three pockets and lay down collar with snap
  • Side Pass throughs to inner clothing
  • Knee length, worn directly over street clothes
  • ARC Rating: 5.7
  • HRC Category: 1
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire.
Specifications
Specifications
Blue
Nomex™ IIIA