Learn More
Sklar InstrumentsSupplier Diversity Partner Worst Pigtail Probe, Sklar, Child size, 5-1/2 in., Sharp, Pointed, Octagonal, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 302461
Worst Pigtail Probe, Sklar, Child size, 5-1/2 in., Sharp, Pointed, Octagonal, Stainless Steel. Sklar's Worst pigtail probe is an ophthalmic tool used to locate a canalicular laceration. The curvature style can bend around the corners of the nasolacrimal duct to the opposing eyelid. This instrument has blunt tips to reduce tissue trauma.
Catalog No. 50-143-8155