Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wortmannin, Penicillium funiculosum, ≥99%

Inhibitor of phosphatidylinositol 3-kinase

$122.40 - $489.00

Specifications

Assay ≥99%
CAS 19545-26-7
Formula Weight 428.44g/mol
Synonym KY 12420
Storage Requirements −30° to −10°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ63983MB View Documents Alfa Aesar™
J63983MB
25mg Each for $264.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ63983MF View Documents Alfa Aesar™
J63983MF
50mg Each for $489.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ63983MA View Documents Alfa Aesar™
J63983MA
10mg Each for $122.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

≥99%
428.44g/mol
−30° to −10°C
Light sensitive
White to Off-white
19545-26-7
KY 12420
Powder
Inhibitor of phosphatidylinositol 3-kinase
Signal Transduction Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit