Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Wortmannin, Penicillium funiculosum, ≥99%

Inhibitor of phosphatidylinositol 3-kinase

Manufacturer:  Alfa Aesar™ J63983MA

 View more versions of this product

Catalog No. AAJ63983MA


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

10mg
−30° to −10°C
Inhibitor of phosphatidylinositol 3-kinase
19545-26-7
428.44g/mol
Light Sensitive
White to Off-white
Powder
≥99%
C23H24O8
KY 12420
Signal Transduction Reagent
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit