Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WR 1065
GREENER_CHOICE

Catalog No. p-4784805
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

p53 activator. Also ROS scavenger

Cell-permeable reactive oxygen species scavenger that activates p53 through a JNK-dependent signaling pathway. Activates p21waf-1 and MDM2, represses transcription of Myc and thymidine kinase, and inhibits DNA topoisomerase IIα. Induces cell cycle arrest and exhibits cytoprotective activity towards normal cells but not cancer cells in vivo. Also exhibits broad spectrum antiviral activity in vitro.

Chemical Identifiers

CAS 14653-77-1
Molecular Formula C5H16Cl2N2S

Specifications

Quantity 10 mg
Formula Weight 207.16
Purity >98%
Molecular Weight (g/mol) 207.16
Chemical Name or Material WR 1065
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.