Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Wraptor Nylon Cloth Labels
Ideal for electrical, telecommunication and datacomm identification. Brady™ Wraptor Nylon Cloth Labels are sized to fit standard blank write-on right-to-lnow labels.
Supplier: Brady™ WRAP8499
Description
- Made from premium grade B-946 vinyl
- Sized to fit standard Blank Write-On Right-To-Know Labels
- Correspond to ANSI proposed Z.535 Standard
- Card-mounted for easy removal
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) | |
Acrylic, Permanent, Cold Temperature | |
Nylon Cloth | |
Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking | |
0.0065 in. |
Suggested Ribbon Series: R4300 | |
White | |
Wraptor Label | |
1 x 1.5 in. (W x H) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction