Wright's Stain, Regular Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 936032

Packaging Glass amber
Quantity 1L

 View more versions of this product

Catalog No. 936032

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-56-1,68988-92-1,7558-79-4,7778-77-0,7664-38-2
    CAS Min % 97.0,0.38,0.08,0.04,0.0010
    CAS Max % 100.0,0.39,0.09,0.04,0.0010
    Physical Form Liquid
    Chemical Name or Material Wright's Stain, Regular Formula

    Specifications

    Physical Form Liquid
    Color Clear dark purple
    Quantity 1L
    CAS Min % 0.04,0.0010,97.0,0.38,0.08
    CAS 7664-38-2,67-56-1,68988-92-1,7558-79-4,7778-77-0
    CAS Max % 0.09,0.04,0.0010,100.0,0.39
    Packaging Glass amber
    Chemical Name or Material Wright's Stain, Regular Formula
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit