Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Model 75 Laboratory Shakers

Bench top shakers with swirling action of hand mixing

Manufacturer:  Burrell Scientific 0757750419

 View more versions of this product

Catalog No. 14-400-120


Add to cart

Description

Description

  • Single Speed with Timer
  • Material: Aluminum and steel
  • Max tested Load at 4.5kg (10 lb.)
  • Color: Blue and white

Warranty and Services

One Year

Specifications

Specifications

60 cycles
Laboratory Shaker
Side arm shafts, Clamps, Power cord, Operations manual, Allen Wrench, Lever arm brake
30.8kg
Accommodates 4 Vessels
385 Oscillations/min.
115V
68 lb.
75-AA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit