Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Simethicone Model 85-AA Laboratory Shakers

Bench top shakers with swirling action of hand mixing

Manufacturer:  Burrell Scientific 0757850436

 View more versions of this product

Catalog No. 14-400-133


This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Description

Description

Burrell Scientific redesigned shaking mechanism and supplies Multi-SUREGRIP™ Clamps and Shafts which allow shaker to achieve required performance levels for Simethicone Foaming Solution Test.
  • Single Speed with Timer
  • Material: Aluminum and steel
  • Max tested Load at 4.5kg (10 lb.)
  • Color: Blue and white

Warranty and Services

One Year

Specifications

Specifications

50 cycles
Simethicone Shaker
Side arm shafts, Clamps, Power cord, Operations manual, Allen Wrench, Lever arm brake
30.8kg
Accommodates 4 vessels
300 Oscillations/min.
220V
68 lb.
85-AA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit