Learn More
WRNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 162381AP
Catalog No. 50-173-0818
Add to cart
Description
Description
Werner ft.s syndrome is a rare autosomal recessive disorder characterized by accelerated aging that is caused by defects in the Werner syndrome ATP-dependent helicase gene (WRN). The protein encoded by this gene interacts with the exonuclease-containing N-terminal portion of the Werner protein. This protein has a ubiquitin-binding zinc-finger domain in the N-terminus, an ATPase domain, and two leucine zipper motifs in the C-terminus. It has sequence similarity to replication factor C family proteins and is conserved from E. coli to human. This protein likely accumulates at sites of DNA damage by interacting with polyubiquinated proteins and also binds to DNA polymerase delta and increases the initiation frequency of DNA polymerase delta-mediated DNA synthesis. This protein also interacts with nucleoporins at nuclear pore complexes. Two transcript variants encoding different isoforms have been isolated for this gene.
Specifications
Specifications
WRNIP1
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CG07, Q91XU0, Q96S55
ATPase WRNIP1, RP11 420G6.2, WHIP, WRNIP1
Rabbit
IgG
150 μL
-20°C
Primary
282835, 56897, 78903
Western Blot
Unconjugated
WRNIP1
Liquid
Wrnip1
WRNIP1 Fusion Protein Ag9040
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat