Learn More
WS 3, Tocris
TRPM8 agonist; cooling agent
Manufacturer:  R&D Systems 2927/50
Quantity 50mg
 View more versions of this product
Catalog No. 29-275-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
211.34
WS 3
C13H25NO
N-Ethyl-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
Specifications
50mg
>99%
211.34
WS 3
39711-79-0
C13H25NO
N-Ethyl-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
Safety and Handling
Recommended Storage : Store at Room Temperature