Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WS 3, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
39711-79-0
C13H25NO
211.349
VUNOFAIHSALQQH-UHFFFAOYSA-N
n-ethyl-p-menthane-3-carboxamide,ethyl menthane carboxamide,n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide,ws-3,n-ethyl-4-menthane-3-carboxamide,n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide,cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl,fema no. 3455,ws 3,n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
62907
N-ethyl-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
CCNC(=O)C1CC(CCC1C(C)C)C
This item is not returnable. View return policy
39711-79-0
C13H25NO
211.349
VUNOFAIHSALQQH-UHFFFAOYSA-N
n-ethyl-p-menthane-3-carboxamide,ethyl menthane carboxamide,n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide,ws-3,n-ethyl-4-menthane-3-carboxamide,n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide,cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl,fema no. 3455,ws 3,n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
62907
N-ethyl-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
CCNC(=O)C1CC(CCC1C(C)C)C

TRPM8 agonist; cooling agent

Cooling agent that is an agonist at TRPM8 receptors (EC 50 = 3.7 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 211.34
Percent Purity >99%
Chemical Name or Material WS 3

Recommended Storage : Store at Room Temperature