Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WTAP Mouse anti-Human, HRP, Clone: OTI12G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274870H

Catalog No. NBP274870H


Add to cart

Description

Description

WTAP Monoclonal antibody specifically detects WTAP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

WTAP
OTI12G7
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
WTAP
Recombinant protein fragment corresponding to amino acids 2-106 of human WTAP (NP_004897) produced in E.coli.
Immunogen affinity purified
Cancer, Tumor Suppressors
Primary
9589.0
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms′ tumour 1-associating protein, WT1-associated protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit