Tocris™ WWL 113

Ces3 and Ces1f inhibitor

Manufacturer: Tocris™ 5259/10

 View all options for this product

Catalog No. 52-591-0

  Check Availability
Add to Cart


For Research Use Only


    Description WWL 113
    Molecular Weight 466.53
    Purity >99
    Quantity 10mg
    Product Type WWL 113

    Carboxylesterase Ces3 and Ces1f inhibitor (IC50 values are ~100nM). Also inhibits ABHD6. Exhibits minimal activity against a panel of other serine hydrolases. Promotes lipid storage in adipocytes. Reduces weight gain in diet-induced obese mice and enhances glucose tolerance in diabetic mice. Orally active.

    • Chemical Name: 4'-[[[Methyl[[3-(4-pyridinyl)phenyl]methyl]amino]carbonyl]oxy]-[1,1'-biphenyl]-4-carboxylic acid ethyl ester
    • Formula: C29H26N2O4
    • CAS Number: 947669-86-5
    • Solubility: Soluble to 50mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit