Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WWL 123

α/β-hydrolase domain 6 (ABHD6) inhibitor; antiepileptic

$205.50 - $848.50

Specifications

Chemical Name or Material N-[[1,1′-Biphenyl]-3-ylmethyl)-N-methylcarbamic acid 4′-(aminocarbonyl)[1,1′-biphenyl]-4-yl ester
Molecular Formula C28H24N2O3
CAS 1338574-83-6
Product Type Target: Hydrolase Inhibitors
Formula Weight 436.5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
531710 View Documents Tocris Bioscience™
5317/10
10mg Each for $205.50
Add to cart
 
531750 View Documents Tocris Bioscience™
5317/50
50mg Each for $848.50
Add to cart
 
Description

Description

α/β-hydrolase domain 6 (ABHD6) inhibitor (IC50 = 0.43 μM). Blocks PTZ -induced seizures in healthy mice and spontaneous seizures in R6/2 mice. Antiepileptic activity is independent of CB1 and CB2 receptors, but is GABAA receptor dependent. Brain penetrant.
Specifications

Specifications

N-[[1,1′-Biphenyl]-3-ylmethyl)-N-methylcarbamic acid 4′-(aminocarbonyl)[1,1′-biphenyl]-4-yl ester
1338574-83-6
436.5
C28H24N2O3
Target: Hydrolase Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit