Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWOX3 Rabbit anti-Human, Non-human Primate, Rat, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc WWOX3301AP

Description
WWOX3 Polyclonal antibody specifically detects WWOX3 in Human, Non-human Primate, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
WWOX3 | |
Polyclonal | |
Unconjugated | |
Wwox | |
WW domain-containing oxidoreductase; WWOX | |
Synthetic peptide taken within amino acid region 250-300 on human WWOXdelta5-8 protein. | |
200 μL | |
Primary | |
Human, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q9NZC7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
292041, 51741, 737907 | |
-20°C | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title