Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310564100UL
Description
WWP2 Polyclonal specifically detects WWP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
WWP2 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
The immunogen is a synthetic peptide directed towards the middle region of human WWP2 (NP_955456). Peptide sequence EMKYTSEGVRYFVDHNTRTTTFKDPRPGFESGTKQGSPGAYDRSFRWKYH | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
11060 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction