Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WY 14643, Tocris Bioscience™

Catalog Number p-4785786
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
50892-23-4
C14H14ClN3O2S
323.80
MFCD00191335
SZRPDCCEHVWOJX-UHFFFAOYSA-N
pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio
5694
CHEBI:32509
2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid
CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C
This item is not returnable. View return policy
50892-23-4
C14H14ClN3O2S
323.80
MFCD00191335
SZRPDCCEHVWOJX-UHFFFAOYSA-N
pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio
5694
CHEBI:32509
2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid
CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C

Selective PPARα agonist

Selective PPARα agonist (EC50 values are 0.63, 32 and > 100 μM at PPARα, PPARγ and PPARδ respectively). Negatively inhibits NF-κB transcriptional activity and decreases the inflammatory response in vitro and in vivo.
Quantity 10 mg
Formula Weight 323.8
Percent Purity >98%
Chemical Name or Material WY 14643
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit