Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WY 45233 succinate, Tocris Bioscience™

Catalog Number 419450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
448904-47-0
C20H31NO6
381.469
ORUUBRMVQCKYHB-XFULWGLBSA-N
unii-h9e1t0bi90 component
76958757
butanedioic acid;4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O.C(CC(=O)O)C(=O)O
This item is not returnable. View return policy
448904-47-0
C20H31NO6
381.469
ORUUBRMVQCKYHB-XFULWGLBSA-N
unii-h9e1t0bi90 component
76958757
butanedioic acid;4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O.C(CC(=O)O)C(=O)O

Dual serotonin/noradrenalin re-uptake inhibitor

Serotonin and noradrenalin reuptake inhibitor (SNRI); selective for human serotonin (SERT) and noradrenalin (NET) transporters against 96 other targets (Ki values are 40.2 and 558.4 nM for SERT and NET respectively). Inhibits [3H]5-HT and [3H]NE uptake (IC50 values are 47.3 and 531.3 nM). Salt form of the major active metabolite of venlafaxine (Cat. No. 2917).
Quantity 50 mg
Formula Weight 381.46
Percent Purity >99%
Chemical Name or Material WY 45233 succinate
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit