X-Gal, OmniPur™, Calbiochem™, EMD Millipore

Chromogenic substrate used in conjunction with IPTG

Manufacturer: EMD Millipore 96601GM

Quantity 1g

 View more versions of this product

Catalog No. 96-601-GM

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7240-90-6
    Physical Form Powder
    Molecular Formula C14H15BrClNO6
    Formula Weight 408.63
    Color White
    Chemical Name or Material X-Gel
    Assay ≥99.0%
    Grade Molecular biology grade

    Specifications

    Formula Weight 408.63
    Quantity 1g
    CAS 7240-90-6
    Physical Form Powder
    Color White
    Molecular Formula C14H15BrClNO6
    Solubility Information Water/DMF/IPTG/: 20mg/mL soluble
    Additional Information Use 4:1 (X-Gal:IPTG)
    Chemical Name or Material X-Gel
    Assay ≥99.0%
    Grade Molecular biology grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit