Learn More
Thermo Scientific™ X-Gal Promotion
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
Manufacturer:  Thermo Scientific™ R0402
 View more versions of this product
Catalog No. FERR0402PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.
Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22µm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
Specifications
Specifications
formulated for direct use in conjunction with IPTG for blue/white colony screening.
X-Gal
2g