Learn More
Corning™ X-MINI™ Pressor
Corning™'s X-MINI™ Pressor allows for the degassing of microbubble-targeted cells. It accommodates a capped 10 mL syringe. The pressure generated using the X-MINI™ Pressor is sufficient to isolate up to 1 x 107 cells, and one syringe can process up to 3 mL of cell suspension at a time.
Supplier:  Corning™ 6915
Catalog No. 10-320-143
Explore available promotions
Add to Cart
Description
Description
Corning™'s X-SERIES™ Systems are manufactured for Corning™ by ThermoGenesis Corp. X-SERIES™, X-WASH™, X-LAB™, X-BACS™, and X-MINI™ are either registered trademarks or trademarks of ThermoGenesis Corp. in the United States and/or other countries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bioprocessing Applications
Corning X-SERIES System
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.