Learn More
Aviva Systems Biology Corp XAB2 Antibody - N-terminal region (ARP50636_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XAB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human XAB2. Alternate names: DKFZp762C1015, HCNP, HCRN, NTC90, SYF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60394