Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAGE1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132314
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA132314 100 μL
1 options

Catalog No. PIPA132314

Supplier: Thermo Scientific PA132314

Rabbit Polyclonal Antibody

PA1-32314 detects XAGE1 from human samples. PA1-32314 has been successfully used in immunoprecipitation, Western blot applications. The PA1-32314 immunogen is: Synthetic peptide made to an internal sequence within the human XAGE-1 protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing′s sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to alternative transcription start sites, results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAGE1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene XAGE1B
Gene Accession No. Q9HD64
Gene Alias CT12.1A, CTP9, GAGED2, XAGE1
Gene Symbols XAGE1B
Host Species Rabbit
Immunogen Synthetic peptide made to an internal sequence within the human XAGE-1 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653067
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.