Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ XAGE2 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP246807PEP
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human XAGE2 The XAGE2 Recombinant Protein Antigen is derived from E. coli. The XAGE2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
PTPDQKREDDQGAAEIQVPDLEADLQ | |
XAGE2 Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4 | |
CT12.2GAGED3, G antigen family D member 3 | |
Unlabeled | |
Recombinant Protein Antigen | |
RUO | |
Human |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20°C short term. Avoid freeze-thaw cycles. | |
AC | |
XAGE2 | |
21 kDa | |
0.1mL | |
E.coli |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction