Learn More
Xaliproden hydrochloride, Tocris
Orally active, high affinity 5-HT1A agonist
Manufacturer:  R&D Systems 2491/50
Quantity 50mg
 View more versions of this product
Catalog No. 24-915-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
417.9
Xaliproden hydrochloride
C24H22F3N·HCl
1,2,3,6-Tetrahydro-1-[2-(2-naphthalenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]-pyridine hydrochloride
Specifications
90494-79-4
C24H22F3N·HCl
417.9
Xaliproden hydrochloride
>99%
50mg
1,2,3,6-Tetrahydro-1-[2-(2-naphthalenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]-pyridine hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature