Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthan Gum, NF, 91-108%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
11138-66-2
C8H14Cl2N2O2
241.11
VXYWXJXCQSDNHX-UHFFFAOYSA-N
2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride
Cl.Cl.NC1=CC(N)=C(OCCO)C=C1
11138-66-2
C8H14Cl2N2O2
241.11
VXYWXJXCQSDNHX-UHFFFAOYSA-N
2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride
Cl.Cl.NC1=CC(N)=C(OCCO)C=C1

XA105, 11138-66-2

Spectrum™ Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Loss on Drying 0.15
Quantity 500 g
CAS Max % 1
Percent Purity 91 to 108%
Grade NF