Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthene 98% Acros Organics

Click to view available options
Quantity:
25g
92-83-1
C13H10O
182.222
MFCD00005055
GJCOSYZMQJWQCA-UHFFFAOYSA-N
xanthene,xanthan,10h-9-oxaanthracene,dibenzo a,e pyran,dibenzopyran, tricyclic,9h-xanthen,xanthane,9h-xanthen-9-yl,acmc-209rhu
7107
CHEBI:10057
9H-xanthene
C1C2=CC=CC=C2OC3=CC=CC=C31
92-83-1
C13H10O
182.222
MFCD00005055
GJCOSYZMQJWQCA-UHFFFAOYSA-N
xanthene,xanthan,10h-9-oxaanthracene,dibenzo a,e pyran,dibenzopyran, tricyclic,9h-xanthen,xanthane,9h-xanthen-9-yl,acmc-209rhu
7107
CHEBI:10057
9H-xanthene
C1C2=CC=CC=C2OC3=CC=CC=C31
Quantity 25g
Color White to Yellow
Melting Point 100°C to 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 17, 73
Formula Weight 182.22
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Xanthene

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.