Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthine monosodium salt monohydrate, ≥96% (dry basis), Affymetrix/USB™

Click to view available options
Quantity:
5g
10g
100g
Packaging:
Plastic bottle
1196-43-6
C5H3N4NaO2
174.095
MFCD00069669
HKDXLLDCXWUUKY-UHFFFAOYSA-M
xanthine sodium salt,xanthine, sodium salt,xanthate de sodium french,1h-purine-2,6-dione, 3,7-dihydro-, sodium salt,3,7-dihydro-1h-purine-2,6-dione sodium salt,3,7-dihydro-1h-purine-2,6-dione, sodium salt,xanthate de sodium,1h-purine-2,6-dione, 3,9-dihydro-, sodium salt 1:?,potassium 2-oxo-3,7-dihydropurin-6-olate,sodium 2-oxo-3,7-dihydropurin-6-olate
23668765
sodium;2-oxo-3,7-dihydropurin-6-olate
C1=NC2=C(N1)C(=NC(=O)N2)[O-].[Na+]
1196-43-6
C5H3N4NaO2
174.095
MFCD00069669
HKDXLLDCXWUUKY-UHFFFAOYSA-M
xanthine sodium salt,xanthine, sodium salt,xanthate de sodium french,1h-purine-2,6-dione, 3,7-dihydro-, sodium salt,3,7-dihydro-1h-purine-2,6-dione sodium salt,3,7-dihydro-1h-purine-2,6-dione, sodium salt,xanthate de sodium,1h-purine-2,6-dione, 3,9-dihydro-, sodium salt 1:?,potassium 2-oxo-3,7-dihydropurin-6-olate,sodium 2-oxo-3,7-dihydropurin-6-olate
23668765
sodium;2-oxo-3,7-dihydropurin-6-olate
C1=NC2=C(N1)C(=NC(=O)N2)[O-].[Na+]
Formula Weight 192.11
Physical Form Crystalline Powder
Percent Purity ≥96%
Packaging Plastic bottle
Quantity 10g
Chemical Name or Material Xanthine Monosodium Salt Monohydrate
Assay Dry basis