Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthurenic acid, Tocris Bioscience™

Catalog Number 412050
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
59-00-7
C10H7NO4
205.17
MFCD00006754
FBZONXHGGPHHIY-UHFFFAOYSA-N
xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
5699
CHEBI:10072
8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1
This item is not returnable. View return policy
59-00-7
C10H7NO4
205.17
MFCD00006754
FBZONXHGGPHHIY-UHFFFAOYSA-N
xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
5699
CHEBI:10072
8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1

Selectively activates group II mGlu receptors

Shown to selectively activate group II mGlu receptors in transfected HEK293 cells at nanomolar concentrations. Attenuates cAMP formation in mouse cortical slices expressing mGlu2 and mGlu3 receptors.
Quantity 50 mg
Formula Weight 205.17
Percent Purity >98%
Chemical Name or Material Xanthurenic acid
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit