Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XCL1/Lymphotactin Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0604-4G24, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212004AF750
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot.Specifications
XCL1/Lymphotactin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
XCL1 | |
Human recombinant XCL1/Lymphotactin | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0604-4G24 | |
Western Blot | |
ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
Mouse | |
0.1 mL | |
6375 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title