Sino Biological™ XCL1 Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human XCL1 (Met 1- Gly 114) fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 10672H08B25

 View more versions of this product

Catalog No. 10-672-H08B25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 11.6kd
    Purity >85% by SDS-PAGE
    Quantity 5 x 5μg
    Source Human
    Common Name Chemokine (C Motif) Ligand 1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Cytokines and Receptors
    Product Type XCL1 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 6375
    Endotoxin Concentration <1 EU/μg
    Gene Symbol XCL1
    Expression System Insect

    Expressed from DNA sequence encoding human XCL1 (Met 1- Gly 114) fused to polyhistidine tag at C-terminus

    • Lyophilized in 20mM, Tris 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 10% glycerol

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit