Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarlite XD Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Smooth, Extra Light and Extra Delicate (SklarLite XD), Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 232225
Sklarlite XD Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Smooth, Extra Light and Extra Delicate (SklarLite XD), Finger Rings, Stainless Steel. Sklarlite XD Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklarlite XD instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with smooth tips and a length of 5-1/4 inches.
Catalog No. 50-143-7970