Learn More
Tocris Bioscience™ XEN 445
Selective endothelial lipase inhibitor; orally bioavailable
$146.90 - $606.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-391-0 View Documents Tocris Bioscience™
5839/10
Each for $146.90
Add to cart
58-395-0 View Documents Tocris Bioscience™
5839/50
Each for $606.00
Add to cart
Description
Description
Selective endothelial lipase (EL) inhibitor (IC50 = 237 nM for human EL). Exhibits >20-fold selectivity for EL over lipoprotein lipase (LPL) and hepatic lipase (HL). Increases high density lipoprotein cholesterol (HDLc) plasma levels in mice. Orally bioavailable.