Learn More
Cytiva XK Chromatography Columns (Empty) PROMO
Ensure trouble-free operation for low to medium pressure. Cytiva XK Chromatography Columns (Empty) connect directly to ÄKTA design instruments or other liquid chromatography systems. The user-friendly ensures reproducible, accurate packing.
$563.40 - $682.20
Specifications
For Use With (Application) Liquid chromatography at low to medium pressure
Particle Size 10 μm pore size
Material Borosilicate glass
Support Type Polypropylene (reinforced glass fiber)/Polyamide Net
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
45-200-47LR1
View Documents
Cytiva
29648095
16 mm
Each for $563.40
Add to cart
45-200-48LR1
View Documents
Cytiva
29648096
26 mm
Each for $682.20
Add to cart
Description
Description
XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.
Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.
  • Plunger design to ensure a homogeneous buffer distribution and reproducible packing
  • Adapter QuickLock mechanism that facilitates column handling and cleaning
  • Fittings for direct connection to chromatographiy systems
  • Cooling jacket for sensitive purifications
  • Extensive range of spare parts available, facilitating maintenance
Specifications
Specifications
Liquid chromatography at low to medium pressure
10 μm pore size
Borosilicate glass
Polypropylene (reinforced glass fiber)/Polyamide Net
SDS
Product Certifications
Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.
Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.