Learn More
Azzota Corp XL-184 DFS Item
Manufacturer:  Azzota Corp C1333
XL-184 (Cabozantinib) is an orally bioavailable inhibitor of c-MET, VEGFR2 and RET. Please request if need more quantities
Catalog No. 50-161-3194