Learn More
Selleck Chemical LLCSupplier Diversity Partner Cabozantinib (BMS-907351) 10mg 849217-68-1 XL184  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S111910MG
Cabozantinib (XL184, BMS-907351) is a potent VEGFR2 inhibitor with IC50 of 0.035 nM and also inhibits c-Met, Ret, Kit, Flt-1/3/4, Tie2, and AXL with IC50 of 1.3 nM, 4 nM, 4.6 nM, 12 nM/11.3 nM/6 nM, 14.3 nM and 7 nM in cell-free assays, respectively. Cabozantinib induces PUMA-dependent apoptosis in colon cancer cells via AKT/GSK-3β/NF-κB signaling pathway.
Catalog No. 50-622-5